Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTATGTTATTTTCCCAGCAACATT[G/T]TGTGATTTTCCAAAAATAAACCATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 134371 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFHR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CFHR1 - complement factor H related 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002113.2 | 180 | Silent Mutation | TTG,TTT | L,F 22 | NP_002104.2 | |
XM_011509457.2 | 180 | Intron | XP_011507759.1 |