Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGCTCTTGCTCAGTCCGCCCTT[C/T]GGGGTGGGGATCTTGCTTCCGGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616690 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP104 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CEP104 - centrosomal protein 104 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014704.3 | 3191 | Silent Mutation | CCA,CCG | P,P 910 | NP_055519.1 | |
XM_005244815.4 | 3191 | Silent Mutation | CCA,CCG | P,P 946 | XP_005244872.1 | |
XM_011542473.2 | 3191 | Silent Mutation | CCA,CCG | P,P 952 | XP_011540775.1 | |
XM_011542474.2 | 3191 | Silent Mutation | CCA,CCG | P,P 916 | XP_011540776.1 | |
XM_011542475.2 | 3191 | Silent Mutation | CCA,CCG | P,P 894 | XP_011540777.1 | |
XM_011542476.2 | 3191 | Silent Mutation | CCA,CCG | P,P 886 | XP_011540778.1 | |
XM_011542477.2 | 3191 | Silent Mutation | CCA,CCG | P,P 828 | XP_011540779.1 | |
XM_011542478.1 | 3191 | Intron | XP_011540780.1 | |||
XM_017002917.1 | 3191 | Silent Mutation | CCA,CCG | P,P 888 | XP_016858406.1 | |
XM_017002918.1 | 3191 | Silent Mutation | CCA,CCG | P,P 852 | XP_016858407.1 | |
XM_017002919.1 | 3191 | Silent Mutation | CCA,CCG | P,P 844 | XP_016858408.1 |