Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCGCATCATCATAGGAGTTGTTA[C/T]CTGGTTTTCCAGAAAAATAGTGAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 139370 MIM: 139340 MIM: 611189 | ||||||||||||||||||||
Literature Links: |
GNAI3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GNAI3 - G protein subunit alpha i3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GNAT2 - G protein subunit alpha transducin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005272.3 | 1088 | Missense Mutation | GAT,GGT | D,G 292 | NP_005263.1 | |
XM_011541264.2 | 1088 | Missense Mutation | GAT,GGT | D,G 292 | XP_011539566.1 |
MIR197 - microRNA 197 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |