Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTCAAAGATTAATGTCCAAGAAG[A/G]CCAGTACCTGGGAAAAGAAACACAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 600278 | ||||||||||||||||||||
Literature Links: |
RAP1GAP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RAP1GAP - RAP1 GTPase activating protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP48 - ubiquitin specific peptidase 48 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032730.1 | 3891 | Intron | NP_001027902.1 | |||
XM_005246009.2 | 3891 | Silent Mutation | GGC,GGT | G,G 979 | XP_005246066.1 | |
XM_006710951.2 | 3891 | Silent Mutation | GGC,GGT | G,G 1029 | XP_006711014.2 | |
XM_006710955.3 | 3891 | Silent Mutation | GGC,GGT | G,G 965 | XP_006711018.1 | |
XM_011542262.2 | 3891 | Silent Mutation | GGC,GGT | G,G 1030 | XP_011540564.1 | |
XM_011542263.2 | 3891 | Silent Mutation | GGC,GGT | G,G 1030 | XP_011540565.1 | |
XM_011542264.1 | 3891 | Silent Mutation | GGC,GGT | G,G 964 | XP_011540566.1 | |
XM_011542265.2 | 3891 | Silent Mutation | GGC,GGT | G,G 904 | XP_011540567.1 | |
XM_011542267.2 | 3891 | Silent Mutation | GGC,GGT | G,G 617 | XP_011540569.1 | |
XM_011542268.2 | 3891 | Silent Mutation | GGC,GGT | G,G 617 | XP_011540570.1 | |
XM_017002495.1 | 3891 | Silent Mutation | GGC,GGT | G,G 964 | XP_016857984.1 | |
XM_017002496.1 | 3891 | Intron | XP_016857985.1 | |||
XM_017002497.1 | 3891 | Silent Mutation | GGC,GGT | G,G 617 | XP_016857986.1 |