Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTTTCAGATGCTGTAGCTTCTT[C/T]CTCCTCTTTCTGCCTCCTTTTCTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 608862 | ||||||||||||||||||||
Literature Links: |
GPATCH4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPATCH4 - G-patch domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015590.3 | 702 | Missense Mutation | AAA,GAA | K,E 220 | NP_056405.2 | |
NM_182679.2 | 702 | Missense Mutation | AAA,GAA | K,E 215 | NP_872620.1 | |
XM_005245287.3 | 702 | Missense Mutation | AAA,GAA | K,E 186 | XP_005245344.1 |
HAPLN2 - hyaluronan and proteoglycan link protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NAXE - NAD(P)HX epimerase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144772.2 | 702 | Intron | NP_658985.2 | |||
XM_017000319.1 | 702 | Intron | XP_016855808.1 |
TTC24 - tetratricopeptide repeat domain 24 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |