Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCCTCTCTGAACCGCAGCAGCTC[C/T]GGGTGGTCGCTGTGCAGGTGGAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607215 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR4689 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR4689 - microRNA 4689 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NPHP4 - nephronophthisis 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291593.1 | 3412 | Silent Mutation | CCA,CCG | P,P 856 | NP_001278522.1 | |
NM_001291594.1 | 3412 | Silent Mutation | CCA,CCG | P,P 857 | NP_001278523.1 | |
NM_015102.4 | 3412 | Silent Mutation | CCA,CCG | P,P 1369 | NP_055917.1 | |
XM_006710563.3 | 3412 | Silent Mutation | CCA,CCG | P,P 1369 | XP_006710626.1 | |
XM_011541213.1 | 3412 | Silent Mutation | CCA,CCG | P,P 1368 | XP_011539515.1 | |
XM_011541214.1 | 3412 | Silent Mutation | CCA,CCG | P,P 1355 | XP_011539516.1 | |
XM_011541215.1 | 3412 | Silent Mutation | CCA,CCG | P,P 1332 | XP_011539517.1 | |
XM_011541216.2 | 3412 | Silent Mutation | CCA,CCG | P,P 1369 | XP_011539518.1 | |
XM_011541217.2 | 3412 | Silent Mutation | CCA,CCG | P,P 1369 | XP_011539519.1 | |
XM_011541218.2 | 3412 | Silent Mutation | CCA,CCG | P,P 1369 | XP_011539520.1 | |
XM_017000996.1 | 3412 | Silent Mutation | CCA,CCG | P,P 1354 | XP_016856485.1 | |
XM_017000997.1 | 3412 | Silent Mutation | CCA,CCG | P,P 1369 | XP_016856486.1 | |
XM_017000998.1 | 3412 | Intron | XP_016856487.1 | |||
XM_017000999.1 | 3412 | Silent Mutation | CCA,CCG | P,P 1193 | XP_016856488.1 | |
XM_017001000.1 | 3412 | Silent Mutation | CCA,CCG | P,P 1193 | XP_016856489.1 | |
XM_017001001.1 | 3412 | Silent Mutation | CCA,CCG | P,P 1103 | XP_016856490.1 | |
XM_017001002.1 | 3412 | Intron | XP_016856491.1 | |||
XM_017001003.1 | 3412 | Silent Mutation | CCA,CCG | P,P 856 | XP_016856492.1 |