Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGCCACAGCAGCTGGAGCCCCCC[A/G]AGGGCTGGCTGCAGCAGCCAGAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612604 MIM: 612605 MIM: 612606 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LCE1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LCE1B - late cornified envelope 1B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LCE1C - late cornified envelope 1C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276331.1 | 260 | Missense Mutation | TCG,TTG | S,L 70 | NP_001263260.1 | |
NM_178351.3 | 260 | Missense Mutation | TCG,TTG | S,L 100 | NP_848128.1 |
LCE1D - late cornified envelope 1D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |