Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGCTGACCCCAGCCTTCCACAAT[G/T]ACATCCTGAAGCCATACGTGGGGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 615341 | ||||||||||||||||||||
Literature Links: |
CYP4A22 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CYP4A22 - cytochrome P450 family 4 subfamily A member 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010969.3 | Intron | NP_001010969.2 | ||||
NM_001308102.1 | Intron | NP_001295031.1 | ||||
XM_005270767.3 | Intron | XP_005270824.1 | ||||
XM_005270770.3 | Intron | XP_005270827.1 |