Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAGGCACTCTTGTCTTCATTTCT[A/G]TTCGATACAGTCAACTCCCAAGGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 131390 | ||||||||||||||||||||
Literature Links: |
NID1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NID1 - nidogen 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002508.2 | 3838 | Nonsense Mutation | CAG,TAG | Q,* 1246 | NP_002499.2 | |
XM_011544195.2 | 3838 | Nonsense Mutation | CAG,TAG | Q,* 1204 | XP_011542497.1 |