Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCGTCCTTTCCAGCACTGGGATT[C/T]GTTGTGGGATCTGGAAGTTGTCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615242 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC27 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC27 - coiled-coil domain containing 27 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRC47 - leucine rich repeat containing 47 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020710.2 | 1636 | Silent Mutation | ACA,ACG | T,T 536 | NP_065761.1 |
SMIM1 - small integral membrane protein 1 (Vel blood group) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |