Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTATTTCTTCTCTAAGTAATCAT[A/G]AATTTTGGCGAAAGGAAATTCATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611540 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SGIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SGIP1 - SH3 domain GRB2 like endophilin interacting protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TCTEX1D1 - Tctex1 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152665.2 | 213 | Missense Mutation | AAA,GAA | K,E 28 | NP_689878.2 | |
XM_011540933.2 | 213 | UTR 5 | XP_011539235.1 | |||
XM_017000566.1 | 213 | Missense Mutation | AAA,GAA | K,E 28 | XP_016856055.1 |