Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTGAGCCACCACAGGTGCTGCAG[G/T]TCCCACCGATGCCGGCGCCAGAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612613 MIM: 612614 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LCE3A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LCE3A - late cornified envelope 3A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LCE3B - late cornified envelope 3B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178433.1 | 186 | Missense Mutation | AGG,AGT | R,S 62 | NP_848520.1 |