Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCAGGAGCCCTAGGTAGCACTCC[A/G]TTTCTTGGTCAGCATGGTTTTAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610640 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
YTHDF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
YTHDF2 - YTH N6-methyladenosine RNA binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172828.1 | 842 | Silent Mutation | CCA,CCG | P,P 65 | NP_001166299.1 | |
NM_001173128.1 | 842 | Silent Mutation | CCA,CCG | P,P 115 | NP_001166599.1 | |
NM_016258.2 | 842 | Silent Mutation | CCA,CCG | P,P 115 | NP_057342.2 |