Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCTCTGTTTTTCAGACCATCGAG[C/T]TAGTGCCTTGAAGTCTCCGGAACAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 603286 | ||||||||||||||||||||
Literature Links: |
GOLT1A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GOLT1A - golgi transport 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198447.1 | 572 | Missense Mutation | AAC,AGC | N,S 129 | NP_940849.1 | |
XM_017000314.1 | 572 | Intron | XP_016855803.1 |
KISS1 - KiSS-1 metastasis-suppressor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |