Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTTTCCAGCTGAGTTATCTGGGT[A/G]TTGTGTCTTTACCTCAGAGTAGACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 605876 | ||||||||||||||||||||
Literature Links: |
FCRL4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FCRL4 - Fc receptor like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031282.2 | 1610 | Missense Mutation | CAC,TAC | H,Y 500 | NP_112572.1 | |
XM_011510034.1 | 1610 | Missense Mutation | CAC,TAC | H,Y 499 | XP_011508336.1 |