Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGCGGCGCCGGGCGCTTGGGGCT[A/G]GAGGAGGCAGCTCGCCTCAGCTGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607813 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLPPR4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLPPR4 - phospholipid phosphatase related 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166252.1 | 227 | Silent Mutation | CTA,CTG | L,L 23 | NP_001159724.1 | |
NM_014839.4 | 227 | Silent Mutation | CTA,CTG | L,L 23 | NP_055654.2 | |
XM_011542498.2 | 227 | Intron | XP_011540800.1 |