Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGATCCAGCTGTACGGGAGCTTGT[A/G]GGAATTGCTCCAGCGACCTGCAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
OLFML2B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OLFML2B - olfactomedin like 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297713.1 | 2710 | Missense Mutation | CAC,TAC | H,Y 558 | NP_001284642.1 | |
NM_015441.2 | 2710 | Missense Mutation | CAC,TAC | H,Y 557 | NP_056256.1 | |
XM_005245075.2 | 2710 | Missense Mutation | CAC,TAC | H,Y 559 | XP_005245132.1 | |
XM_011509398.2 | 2710 | Missense Mutation | CAC,TAC | H,Y 317 | XP_011507700.1 | |
XM_017000967.1 | 2710 | Intron | XP_016856456.1 |