Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGAGTCACTCCTGCTCCGGGAGC[A/G]GCTCTGCGACTTGGACCCACCAGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613482 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCNL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCNL2 - cyclin L2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039577.3 | 1174 | Intron | NP_001034666.1 | |||
NM_001320153.1 | 1174 | Missense Mutation | CGC,TGC | R,C 198 | NP_001307082.1 | |
NM_001320155.1 | 1174 | Missense Mutation | CGC,TGC | R,C 198 | NP_001307084.1 | |
NM_030937.4 | 1174 | Missense Mutation | CGC,TGC | R,C 420 | NP_112199.2 | |
XM_011542216.2 | 1174 | Missense Mutation | CGC,TGC | R,C 453 | XP_011540518.1 | |
XM_011542219.2 | 1174 | Missense Mutation | CGC,TGC | R,C 289 | XP_011540521.1 | |
XM_011542221.2 | 1174 | Missense Mutation | CGC,TGC | R,C 198 | XP_011540523.1 | |
XM_017002420.1 | 1174 | Missense Mutation | CGC,TGC | R,C 298 | XP_016857909.1 | |
XM_017002421.1 | 1174 | Missense Mutation | CGC,TGC | R,C 289 | XP_016857910.1 | |
XM_017002422.1 | 1174 | Missense Mutation | CGC,TGC | R,C 198 | XP_016857911.1 | |
XM_017002423.1 | 1174 | Intron | XP_016857912.1 |