Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCGGCGGAGGAGCGGATGGCAGAG[A/G]AAGGAGGCGGCGGCCAAGGCGACGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609100 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO28 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXO28 - F-box protein 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136115.2 | 77 | Missense Mutation | AAA,GAA | K,E 12 | NP_001129587.1 | |
NM_015176.3 | 77 | Missense Mutation | AAA,GAA | K,E 12 | NP_055991.1 |