Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATTCAGGTGTGGCTCTATGAGCA[A/G]GTGAATATGCGGATAGAAGGCTGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 128260 MIM: 613513 | ||||||||||||||||||||
Literature Links: |
SNRPE PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SNRPE - small nuclear ribonucleoprotein polypeptide E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304464.1 | 189 | Silent Mutation | CAA,CAG | Q,Q 30 | NP_001291393.1 | |
NM_003094.3 | 189 | Silent Mutation | CAA,CAG | Q,Q 38 | NP_003085.1 |
ZC3H11A - zinc finger CCCH-type containing 11A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |