Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTTTGAACAGGCTTGAGCAGTTC[A/G]GCTGTTTGGTTGGCTGTAAGGTTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
PLD5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLD5 - phospholipase D family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195811.1 | 1750 | Missense Mutation | NP_001182740.1 | |||
NM_001195812.1 | 1750 | Missense Mutation | NP_001182741.1 | |||
NM_001320272.1 | 1750 | Missense Mutation | NP_001307201.1 | |||
NM_152666.2 | 1750 | Missense Mutation | NP_689879.2 | |||
XM_006711752.2 | 1750 | Missense Mutation | XP_006711815.1 | |||
XM_011544115.2 | 1750 | Missense Mutation | XP_011542417.1 | |||
XM_011544116.2 | 1750 | Missense Mutation | XP_011542418.1 | |||
XM_011544119.2 | 1750 | Missense Mutation | XP_011542421.2 | |||
XM_011544120.2 | 1750 | Missense Mutation | XP_011542422.1 | |||
XM_011544121.2 | 1750 | Missense Mutation | XP_011542423.1 | |||
XM_011544122.2 | 1750 | Missense Mutation | XP_011542424.1 | |||
XM_017000567.1 | 1750 | Missense Mutation | XP_016856056.1 | |||
XM_017000568.1 | 1750 | Missense Mutation | XP_016856057.1 | |||
XM_017000569.1 | 1750 | Missense Mutation | XP_016856058.1 | |||
XM_017000570.1 | 1750 | Missense Mutation | XP_016856059.1 |