Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTTCAAAAAGGTACTGTACCCCA[C/T]GCTATAGTGAGTTTGAAGAGCTCGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609764 | ||||||||||||||||||||
Literature Links: |
KDM4A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KDM4A - lysine demethylase 4A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014663.2 | 531 | Missense Mutation | CGC,TGC | R,C 110 | NP_055478.2 | |
XM_005271354.3 | 531 | Missense Mutation | CGC,TGC | R,C 110 | XP_005271411.1 | |
XM_005271355.3 | 531 | Missense Mutation | CGC,TGC | R,C 110 | XP_005271412.1 | |
XM_005271356.3 | 531 | Missense Mutation | CGC,TGC | R,C 19 | XP_005271413.1 | |
XM_011542467.2 | 531 | Intron | XP_011540769.2 | |||
XM_017002912.1 | 531 | Missense Mutation | CGC,TGC | R,C 19 | XP_016858401.1 |