Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGCAAGTACTCCGAGATGAACAA[C/T]CAAATTGACATCATGCAGCTGCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601231 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANGPTL7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANGPTL7 - angiopoietin like 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021146.3 | 612 | Silent Mutation | AAC,AAT | N,N 107 | NP_066969.1 | |
XM_017000004.1 | 612 | Silent Mutation | AAC,AAT | N,N 107 | XP_016855493.1 |
MTOR - mechanistic target of rapamycin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004958.3 | 612 | Intron | NP_004949.1 | |||
XM_005263438.2 | 612 | Intron | XP_005263495.1 | |||
XM_011541166.2 | 612 | Intron | XP_011539468.1 | |||
XM_017000900.1 | 612 | Intron | XP_016856389.1 | |||
XM_017000901.1 | 612 | Intron | XP_016856390.1 | |||
XM_017000902.1 | 612 | Intron | XP_016856391.1 |