Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGATTCTGTGCGAGAACGGCTGG[G/T]GGCCAAGCTGGCAGACACCGTGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 608862 | ||||||||||||||||||||
Literature Links: |
NAXE PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NAXE - NAD(P)HX epimerase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTC24 - tetratricopeptide repeat domain 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105669.2 | 1220 | Missense Mutation | GGG,GTG | G,V 395 | NP_001099139.2 | |
XM_011509253.2 | 1220 | Missense Mutation | GGG,GTG | G,V 192 | XP_011507555.1 | |
XM_011509254.2 | 1220 | Missense Mutation | GGG,GTG | G,V 191 | XP_011507556.1 | |
XM_011509255.2 | 1220 | Missense Mutation | GGG,GTG | G,V 192 | XP_011507557.1 | |
XM_017000497.1 | 1220 | Missense Mutation | GGG,GTG | G,V 63 | XP_016855986.1 |