Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTCGAAGCTTCCTGCACATTCCA[A/T]GTGTTTTGGGGGAATTTGTTTTCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CCDC190 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC190 - coiled-coil domain containing 190 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178550.4 | 3484 | Missense Mutation | ATG,TTG | M,L 231 | NP_848645.3 | |
XM_005245125.3 | 3484 | Missense Mutation | ATG,TTG | M,L 242 | XP_005245182.1 | |
XM_006711289.3 | 3484 | Missense Mutation | ATG,TTG | M,L 230 | XP_006711352.1 | |
XM_011509477.2 | 3484 | Missense Mutation | ATG,TTG | M,L 243 | XP_011507779.1 |