Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCCCGCAGCTGGGCAGCGTGAAG[C/G]CTTTTCTTCAGGTCCTTGCACTTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 604043 | ||||||||||||||||||||
Literature Links: |
LOC105748977 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC105748977 - uncharacterized LOC105748977 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NEK2 - NIMA related kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204182.1 | 1372 | Intron | NP_001191111.1 | |||
NM_001204183.1 | 1372 | Intron | NP_001191112.1 | |||
NM_002497.3 | 1372 | Missense Mutation | AGC,AGG | S,R 416 | NP_002488.1 | |
XM_005273147.1 | 1372 | Missense Mutation | AGC,AGG | S,R 408 | XP_005273204.1 |