Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAAGAGGAAAAGGAGGAGCAGAG[C/T]ACAAATTCAACACTTGACCCTGTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM177B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM177B - family with sequence similarity 177 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324080.1 | 944 | Silent Mutation | AGC,AGT | S,S 51 | NP_001311009.1 | |
NM_207468.2 | 944 | Silent Mutation | AGC,AGT | S,S 51 | NP_997351.2 | |
XM_017001279.1 | 944 | Silent Mutation | AGC,AGT | S,S 51 | XP_016856768.1 | |
XM_017001280.1 | 944 | Silent Mutation | AGC,AGT | S,S 51 | XP_016856769.1 | |
XM_017001281.1 | 944 | Silent Mutation | AGC,AGT | S,S 51 | XP_016856770.1 | |
XM_017001282.1 | 944 | Silent Mutation | AGC,AGT | S,S 51 | XP_016856771.1 | |
XM_017001283.1 | 944 | Silent Mutation | AGC,AGT | S,S 62 | XP_016856772.1 | |
XM_017001284.1 | 944 | Silent Mutation | AGC,AGT | S,S 62 | XP_016856773.1 | |
XM_017001285.1 | 944 | Silent Mutation | AGC,AGT | S,S 62 | XP_016856774.1 |