Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATAGGCTTGGACTGAGATTTTCTC[G/T]TTGACTCCATGGATGCTGAGGAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PM20D1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PM20D1 - peptidase M20 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152491.4 | 1302 | Silent Mutation | NP_689704.4 | |||
XM_017000396.1 | 1302 | Intron | XP_016855885.1 | |||
XM_017000397.1 | 1302 | Silent Mutation | XP_016855886.1 | |||
XM_017000398.1 | 1302 | Missense Mutation | XP_016855887.1 |