Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATAGCCAATTCTCAGGATGGATAT[A/G]TATGGCAAGTCACTGACATGAATCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 600063 MIM: 610667 | ||||||||||||||||||||
Literature Links: |
TROVE2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TROVE2 - TROVE domain family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042369.2 | 255 | Missense Mutation | ATA,GTA | I,V 24 | NP_001035828.1 | |
NM_001042370.2 | 255 | Missense Mutation | ATA,GTA | I,V 24 | NP_001035829.2 | |
NM_001173524.1 | 255 | Missense Mutation | ATA,GTA | I,V 24 | NP_001166995.1 | |
NM_001173525.1 | 255 | Missense Mutation | ATA,GTA | I,V 24 | NP_001166996.1 | |
NM_004600.5 | 255 | Missense Mutation | ATA,GTA | I,V 24 | NP_004591.2 | |
XM_006711495.3 | 255 | Missense Mutation | ATA,GTA | I,V 24 | XP_006711558.1 | |
XM_006711496.3 | 255 | Missense Mutation | ATA,GTA | I,V 24 | XP_006711559.1 | |
XM_006711497.3 | 255 | Missense Mutation | ATA,GTA | I,V 24 | XP_006711560.1 | |
XM_011509922.2 | 255 | Intron | XP_011508224.1 | |||
XM_017002180.1 | 255 | Missense Mutation | ATA,GTA | I,V 24 | XP_016857669.1 | |
XM_017002181.1 | 255 | Missense Mutation | ATA,GTA | I,V 24 | XP_016857670.1 | |
XM_017002182.1 | 255 | Missense Mutation | ATA,GTA | I,V 24 | XP_016857671.1 | |
XM_017002183.1 | 255 | Intron | XP_016857672.1 |
UCHL5 - ubiquitin C-terminal hydrolase L5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |