Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCAGGGAATTTGGAGCCACAGTAC[A/G]GGGCCAGTGTCCATCGTGAGACGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 613692 | ||||||||||||||||||||
Literature Links: |
ZCCHC11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZCCHC11 - zinc finger CCHC-type containing 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009881.2 | 4223 | Missense Mutation | CGT,TGT | R,C 1554 | NP_001009881.1 | |
NM_015269.2 | 4223 | Missense Mutation | CGT,TGT | R,C 1553 | NP_056084.1 | |
XM_005270676.2 | 4223 | Missense Mutation | CGT,TGT | R,C 1554 | XP_005270733.1 | |
XM_005270678.2 | 4223 | Missense Mutation | CGT,TGT | R,C 1553 | XP_005270735.1 | |
XM_005270679.2 | 4223 | Missense Mutation | CGT,TGT | R,C 1549 | XP_005270736.1 | |
XM_005270680.2 | 4223 | Missense Mutation | CGT,TGT | R,C 1549 | XP_005270737.1 | |
XM_005270681.2 | 4223 | Missense Mutation | CGT,TGT | R,C 1516 | XP_005270738.1 | |
XM_005270682.2 | 4223 | Missense Mutation | CGT,TGT | R,C 1443 | XP_005270739.1 | |
XM_005270683.2 | 4223 | Missense Mutation | CGT,TGT | R,C 1432 | XP_005270740.1 | |
XM_006710498.2 | 4223 | Missense Mutation | CGT,TGT | R,C 1554 | XP_006710561.1 | |
XM_006710499.3 | 4223 | Missense Mutation | CGT,TGT | R,C 1554 | XP_006710562.1 | |
XM_006710500.1 | 4223 | Missense Mutation | CGT,TGT | R,C 1483 | XP_006710563.1 | |
XM_011541097.1 | 4223 | Missense Mutation | CGT,TGT | R,C 1554 | XP_011539399.1 | |
XM_011541098.1 | 4223 | Missense Mutation | CGT,TGT | R,C 1527 | XP_011539400.1 | |
XM_011541099.1 | 4223 | Missense Mutation | CGT,TGT | R,C 1456 | XP_011539401.1 | |
XM_011541100.2 | 4223 | Intron | XP_011539402.1 | |||
XM_011541101.2 | 4223 | Intron | XP_011539403.1 | |||
XM_017000803.1 | 4223 | Missense Mutation | CGT,TGT | R,C 1522 | XP_016856292.1 | |
XM_017000804.1 | 4223 | Missense Mutation | CGT,TGT | R,C 1522 | XP_016856293.1 | |
XM_017000805.1 | 4223 | Missense Mutation | CGT,TGT | R,C 1511 | XP_016856294.1 | |
XM_017000806.1 | 4223 | Missense Mutation | CGT,TGT | R,C 1478 | XP_016856295.1 | |
XM_017000807.1 | 4223 | Missense Mutation | CGT,TGT | R,C 1478 | XP_016856296.1 | |
XM_017000808.1 | 4223 | Missense Mutation | CGT,TGT | R,C 1438 | XP_016856297.1 | |
XM_017000809.1 | 4223 | Missense Mutation | CGT,TGT | R,C 1443 | XP_016856298.1 | |
XM_017000810.1 | 4223 | Missense Mutation | CGT,TGT | R,C 1438 | XP_016856299.1 | |
XM_017000811.1 | 4223 | Missense Mutation | CGT,TGT | R,C 1432 | XP_016856300.1 | |
XM_017000812.1 | 4223 | Missense Mutation | CGT,TGT | R,C 1431 | XP_016856301.1 | |
XM_017000813.1 | 4223 | Missense Mutation | CGT,TGT | R,C 1427 | XP_016856302.1 | |
XM_017000814.1 | 4223 | Missense Mutation | CGT,TGT | R,C 1321 | XP_016856303.1 | |
XM_017000815.1 | 4223 | Intron | XP_016856304.1 | |||
XM_017000816.1 | 4223 | Intron | XP_016856305.1 | |||
XM_017000817.1 | 4223 | Intron | XP_016856306.1 | |||
XM_017000818.1 | 4223 | Intron | XP_016856307.1 |