Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGTCAAATATATAGAAATGAAGAG[A/G]GTTGCAGAAGCTAAGGTAATTCACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609440 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UTP11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UTP11 - UTP11, small subunit processome component homolog (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016037.3 | 352 | Silent Mutation | AGA,AGG | R,R 109 | NP_057121.2 | |
XM_011541555.2 | 352 | Silent Mutation | AGA,AGG | R,R 40 | XP_011539857.1 |