Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGGAAGCCCCAATCAGTTCTTC[C/T]TCGTTTTGGGAGTGTCATATTTCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 600867 | ||||||||||||||||||||
Literature Links: |
SSR2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SSR2 - signal sequence receptor subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003145.3 | 622 | Missense Mutation | AAG,AGG | K,R 181 | NP_003136.1 | |
XM_011509923.2 | 622 | Missense Mutation | AAG,AGG | K,R 200 | XP_011508225.1 |