Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGGCGCCCTGTCAAATTCTCATC[A/G]AAGTGTCTAAAGGATAAAAAAAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 182860 | ||||||||||||||||||||
Literature Links: |
OR10Z1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OR10Z1 - olfactory receptor family 10 subfamily Z member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPTA1 - spectrin alpha, erythrocytic 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003126.2 | 6944 | Silent Mutation | TTC,TTT | F,F 2283 | NP_003117.2 | |
XM_011509916.2 | 6944 | Silent Mutation | TTC,TTT | F,F 2283 | XP_011508218.1 | |
XM_011509917.2 | 6944 | Silent Mutation | TTC,TTT | F,F 2277 | XP_011508219.1 | |
XM_011509918.2 | 6944 | Intron | XP_011508220.1 | |||
XM_011509919.2 | 6944 | Intron | XP_011508221.1 |