Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACAGCCAAGCTGCAGCTGCAGAC[G/T]CGGCGGCCCTCATATCTGGAGTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153390 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM167B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM167B - family with sequence similarity 167 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032648.2 | 319 | Silent Mutation | ACG,ACT | T,T 38 | NP_116037.2 |
LCK - LCK proto-oncogene, Src family tyrosine kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MTMR9LP - myotubularin related protein 9-like, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |