Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGTACAGGGCTGTCACTGTCCA[C/T]AGGGATGACTGCCACGTCGCAGGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 610171 | ||||||||||||||||||||
Literature Links: |
CALML6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CALML6 - calmodulin like 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005244729.3 | 349 | Intron | XP_005244786.2 | |||
XM_011540841.2 | 349 | Intron | XP_011539143.1 |
CFAP74 - cilia and flagella associated protein 74 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM52 - transmembrane protein 52 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178545.3 | 349 | Missense Mutation | ATG,GTG | M,V 105 | NP_848640.1 |