Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCCTTGCCAAAAACATCCCGGAG[C/G]CTTAGGAAAGTGTTGTTCCTTCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 134390 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
F3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
F3 - coagulation factor III, tissue factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178096.1 | 743 | Missense Mutation | AGC,AGG | S,R 174 | NP_001171567.1 | |
NM_001993.4 | 743 | Missense Mutation | AGC,AGG | S,R 174 | NP_001984.1 |