Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCACGGTTAGGCCGACCCATAGG[A/G]GCCCTGGCCAGGGGAGCAATCGGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 614206 MIM: 601989 | ||||||||||||||||||||
Literature Links: |
CHTOP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHTOP - chromatin target of PRMT1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206612.1 | 658 | Silent Mutation | GGA,GGG | G,G 94 | NP_001193541.1 | |
NM_001244664.1 | 658 | Intron | NP_001231593.1 | |||
NM_001317077.1 | 658 | Silent Mutation | GGA,GGG | G,G 93 | NP_001304006.1 | |
NM_015607.3 | 658 | Silent Mutation | GGA,GGG | G,G 93 | NP_056422.2 | |
XM_017000993.1 | 658 | Silent Mutation | GGA,GGG | G,G 94 | XP_016856482.1 |
S100A13 - S100 calcium binding protein A13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |