Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCATCAGAGTTCTTGCTGGAGTT[C/T]CGGCTCTTGAAACGCACCAGAAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 153420 MIM: 603491 | ||||||||||||||||||||
Literature Links: |
CD58 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD58 - CD58 molecule | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IGSF3 - immunoglobulin superfamily member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007237.2 | 4146 | Silent Mutation | CGA,CGG | R,R 1152 | NP_001007238.1 | |
NM_001542.3 | 4146 | Silent Mutation | CGA,CGG | R,R 1172 | NP_001533.2 | |
XM_005270793.2 | 4146 | Silent Mutation | CGA,CGG | R,R 1172 | XP_005270850.1 | |
XM_005270794.4 | 4146 | Silent Mutation | CGA,CGG | R,R 1157 | XP_005270851.1 | |
XM_006710593.3 | 4146 | Silent Mutation | CGA,CGG | R,R 1177 | XP_006710656.1 | |
XM_011541315.1 | 4146 | Silent Mutation | CGA,CGG | R,R 1172 | XP_011539617.1 | |
XM_011541316.2 | 4146 | Silent Mutation | CGA,CGG | R,R 615 | XP_011539618.1 |