Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATGGACAATCACAGTGGTGGTGG[G/T]GGGGGTAGCAGGTAAATTGCTTAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 614601 | ||||||||||||||||||||
Literature Links: |
ZNF326 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF326 - zinc finger protein 326 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320185.1 | 361 | Silent Mutation | GGG,GGT | G,G 66 | NP_001307114.1 | |
NM_181781.3 | 361 | Intron | NP_861446.2 | |||
NM_182975.3 | 361 | Silent Mutation | GGG,GGT | G,G 66 | NP_892020.1 | |
NM_182976.3 | 361 | Silent Mutation | GGG,GGT | G,G 66 | NP_892021.1 | |
XM_005270779.4 | 361 | UTR 5 | XP_005270836.1 | |||
XM_011541290.2 | 361 | UTR 5 | XP_011539592.1 |