Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGCTGTGCTGAAACTTGAGCCCC[C/T]GTGGATCAACGTGCTCCAGGAGGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 146790 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FCGR2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FCGR2A - Fc fragment of IgG receptor IIa | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136219.1 | 181 | Missense Mutation | CCG,CTG | P,L 48 | NP_001129691.1 | |
NM_021642.3 | 181 | Missense Mutation | CCG,CTG | P,L 47 | NP_067674.2 | |
XM_011509287.2 | 181 | Missense Mutation | CCG,CTG | P,L 48 | XP_011507589.1 | |
XM_011509290.2 | 181 | Missense Mutation | CCG,CTG | P,L 48 | XP_011507592.1 | |
XM_011509291.1 | 181 | Missense Mutation | CCG,CTG | P,L 48 | XP_011507593.1 | |
XM_017000663.1 | 181 | Missense Mutation | CCG,CTG | P,L 47 | XP_016856152.1 | |
XM_017000664.1 | 181 | Missense Mutation | CCG,CTG | P,L 48 | XP_016856153.1 | |
XM_017000665.1 | 181 | Missense Mutation | CCG,CTG | P,L 48 | XP_016856154.1 | |
XM_017000666.1 | 181 | Missense Mutation | CCG,CTG | P,L 48 | XP_016856155.1 | |
XM_017000667.1 | 181 | Missense Mutation | CCG,CTG | P,L 48 | XP_016856156.1 | |
XM_017000668.1 | 181 | Missense Mutation | CCG,CTG | P,L 47 | XP_016856157.1 |