Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCGACAGCAGATGCAGGACAGGAA[G/T]GCTGAGGACTCTTTCTGATACTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607399 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MCOLN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MCOLN2 - mucolipin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153259.3 | 1719 | Silent Mutation | GCA,GCC | A,A 545 | NP_694991.2 | |
XM_005270719.3 | 1719 | Silent Mutation | GCA,GCC | A,A 517 | XP_005270776.1 | |
XM_006710552.2 | 1719 | Silent Mutation | GCA,GCC | A,A 517 | XP_006710615.1 | |
XM_011541187.2 | 1719 | Silent Mutation | GCA,GCC | A,A 517 | XP_011539489.1 | |
XM_011541188.2 | 1719 | Silent Mutation | GCA,GCC | A,A 517 | XP_011539490.1 | |
XM_017000923.1 | 1719 | Intron | XP_016856412.1 |