Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAAGGACCGTGGCTCTGTCTCTG[A/G]CCCAGAGGCCTCCCGCCAGCGCTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611315 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZSCAN20 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZSCAN20 - zinc finger and SCAN domain containing 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145238.3 | 290 | Missense Mutation | GAC,GGC | D,G 46 | NP_660281.2 | |
XM_005271171.3 | 290 | Missense Mutation | GAC,GGC | D,G 46 | XP_005271228.1 | |
XM_006710874.3 | 290 | Missense Mutation | GAC,GGC | D,G 46 | XP_006710937.1 | |
XM_006710876.3 | 290 | Missense Mutation | GAC,GGC | D,G 46 | XP_006710939.1 | |
XM_017002237.1 | 290 | Missense Mutation | GAC,GGC | D,G 46 | XP_016857726.1 | |
XM_017002238.1 | 290 | Missense Mutation | GAC,GGC | D,G 46 | XP_016857727.1 | |
XM_017002239.1 | 290 | UTR 5 | XP_016857728.1 | |||
XM_017002240.1 | 290 | UTR 5 | XP_016857729.1 | |||
XM_017002241.1 | 290 | Intron | XP_016857730.1 |