Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACACCCACCTTTGCAGTAAATCTC[A/G]CCATCCTTGTCTGCCAGGGTGGTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 123876 | ||||||||||||||||||||
Literature Links: |
CSRP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CSRP1 - cysteine and glycine rich protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193570.1 | 706 | Silent Mutation | GGC,GGT | G,G 157 | NP_001180499.1 | |
NM_001193571.1 | 706 | Silent Mutation | GGC,GGT | G,G 163 | NP_001180500.1 | |
NM_001193572.1 | 706 | Silent Mutation | GGC,GGT | G,G 163 | NP_001180501.1 | |
NM_004078.2 | 706 | Silent Mutation | GGC,GGT | G,G 163 | NP_004069.1 |