Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAGTTGAAGGACCTGTTGGCGTT[A/G]TCTCCTGTTCTTCTTTCTGATTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TDRD5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TDRD5 - tudor domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199085.1 | 953 | Silent Mutation | TTA,TTG | L,L 145 | NP_001186014.1 | |
NM_001199089.1 | 953 | Silent Mutation | TTA,TTG | L,L 145 | NP_001186018.1 | |
NM_001199091.1 | 953 | Silent Mutation | TTA,TTG | L,L 145 | NP_001186020.1 | |
NM_001199092.1 | 953 | Intron | NP_001186021.1 | |||
NM_173533.3 | 953 | Silent Mutation | TTA,TTG | L,L 145 | NP_775804.2 | |
XM_005244934.1 | 953 | Silent Mutation | TTA,TTG | L,L 145 | XP_005244991.1 | |
XM_005244935.4 | 953 | Silent Mutation | TTA,TTG | L,L 145 | XP_005244992.1 | |
XM_017000473.1 | 953 | Silent Mutation | TTA,TTG | L,L 145 | XP_016855962.1 |