Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGTGAGCCTCCTGCTGCAGGTGC[C/T]CAATGTGGGAGGCGGCTTGGAGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604630 MIM: 610325 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NR0B2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NR0B2 - nuclear receptor subfamily 0 group B member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021969.2 | 702 | Missense Mutation | GAG,GGG | E,G 189 | NP_068804.1 | |
XM_011542297.2 | 702 | Intron | XP_011540599.1 |
NUDC - nuclear distribution C, dynein complex regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006600.3 | 702 | Intron | NP_006591.1 | |||
XM_011540529.1 | 702 | Intron | XP_011538831.1 | |||
XM_011540530.1 | 702 | Intron | XP_011538832.1 | |||
XM_017000094.1 | 702 | Intron | XP_016855583.1 | |||
XM_017000095.1 | 702 | Intron | XP_016855584.1 | |||
XM_017000096.1 | 702 | Intron | XP_016855585.1 |