Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCATCTTTTCTGTGATGAACTT[G/T]TTCTTTCCGAGACATTTGCTGGGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 116951 | ||||||||||||||||||||
Literature Links: |
CDK11A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDK11A - cyclin dependent kinase 11A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001313896.1 | 607 | Missense Mutation | AAA,CAA | K,Q 96 | NP_001300825.1 | |
NM_001313982.1 | 607 | Missense Mutation | AAA,CAA | K,Q 96 | NP_001300911.1 | |
NM_024011.3 | 607 | Missense Mutation | AAA,CAA | K,Q 96 | NP_076916.2 | |
NM_033529.3 | 607 | Missense Mutation | AAA,CAA | K,Q 96 | NP_277071.2 |
SLC35E2 - solute carrier family 35 member E2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |