Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCCAGCATCTGCTTCTGTTCCTG[A/G]GGGGGTGCTGCAGCCAGCATGGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603407 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929516 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101929516 - uncharacterized LOC101929516 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PABPC4 - poly(A) binding protein cytoplasmic 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135653.1 | Intron | NP_001129125.1 | ||||
NM_001135654.1 | Intron | NP_001129126.1 | ||||
NM_003819.3 | Intron | NP_003810.1 |
PPIEL - peptidylprolyl isomerase E like pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA55 - small nucleolar RNA, H/ACA box 55 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |