Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCGGACATGGATAACGTTTCTAC[A/G]CGGCAGATGCTTTCAACGTAGACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612866 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALG14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALG14 - ALG14, UDP-N-acetylglucosaminyltransferase subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305242.1 | 584 | UTR 3 | NP_001292171.1 | |||
NM_144988.3 | 584 | Silent Mutation | CGC,CGT | R,R 177 | NP_659425.1 | |
XM_005270582.3 | 584 | Intron | XP_005270639.1 | |||
XM_011540897.2 | 584 | Intron | XP_011539199.1 |