Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAACCGAGGAAATGAAGACGCGAA[A/G]GACTACCCGCCTTCAGCAGCAGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614512 MIM: 614513 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TOR1AIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TOR1AIP1 - torsin 1A interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267578.1 | 838 | Missense Mutation | AAG,AGG | K,R 126 | NP_001254507.1 | |
NM_015602.3 | 838 | Missense Mutation | AAG,AGG | K,R 126 | NP_056417.2 | |
XM_011509403.1 | 838 | Missense Mutation | AAG,AGG | K,R 126 | XP_011507705.1 | |
XM_011509404.1 | 838 | Missense Mutation | AAG,AGG | K,R 126 | XP_011507706.1 |
TOR1AIP2 - torsin 1A interacting protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |