Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCATCACTGTCTCTGCAGAGGCT[C/T]GGGGTGGTCTGGGGGCCCCTCCGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102771 MIM: 139340 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMPD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AMPD2 - adenosine monophosphate deaminase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257360.1 | 472 | Missense Mutation | CGG,TGG | R,W 87 | NP_001244289.1 | |
NM_001257361.1 | 472 | UTR 5 | NP_001244290.1 | |||
NM_001308170.1 | 472 | Intron | NP_001295099.1 | |||
NM_004037.7 | 472 | Missense Mutation | CGG,TGG | R,W 87 | NP_004028.3 | |
NM_139156.3 | 472 | Missense Mutation | CGG,TGG | R,W 6 | NP_631895.1 | |
NM_203404.1 | 472 | Intron | NP_981949.1 |
GNAT2 - G protein subunit alpha transducin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |